4-(Triethoxysilyl)anisole 97%

Supplier: Ambeed
Warning

Synonyms: Triethoxy(4-methoxyphenyl)silane

A819280-1G A819280-25G A819280-5G
77256-108EA 19.57 USD
77256-108 77256-110 77256-112
4-(Triethoxysilyl)anisole 97%
4-(Triethoxysilyl)anisole

Precaución: for Research Use Only

Formula: C₁₃H₂₂O₄Si
MW: 270.3969 g/mol
MDL Number: MFCD05665142
CAS Number: 21130-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR