(1R)-1-[Bis(1,1-dimethylethyl)phosphino]-2-[(1R)-1-(diphenylphosphino)ethyl]ferrocene 98%

Supplier: Ambeed
Warning

A819215-100MG A819215-1G A819215-250MG
77256-102EA 14 USD
77256-102 77256-104 77256-106
(1R)-1-[Bis(1,1-dimethylethyl)phosphino]-2-[(1R)-1-(diphenylphosphino)ethyl]ferrocene 98%
(1R)-1-[Bis(1,1-dimethylethyl)phosphino]-2-[(1R)-1-(diphenylphosphino)ethyl]ferrocene

Precaución: for Research Use Only

Formula: C₃₂H₄₀FeP₂
MW: 542.45 g/mol
MDL Number: MFCD08561170
CAS Number: 223120-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR