3-(3,4-Dimethoxyphenyl)-1-(3-hydroxyphenyl)propan-1-one 95%

Supplier: Ambeed
Warning

A819174-100MG A819174-1G A819174-250MG A819174-5G
77281-464EA 42.23 USD
77281-464 77281-466 77281-468 77281-470
3-(3,4-Dimethoxyphenyl)-1-(3-hydroxyphenyl)propan-1-one 95%
3-(3,4-Dimethoxyphenyl)-1-(3-hydroxyphenyl)propan-1-one

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₄
MW: 286.322 g/mol
MDL Number: MFCD06245367
CAS Number: 178445-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR