4-(Ethoxymethylene)-2-phenyloxazol-5(4H)-one 95%

Supplier: Ambeed
Warning

Synonyms: Oxazolone, 4-(ethoxymethylene)-2-phenyloxazol-5(4H)-one

A818543-1G A818543-100MG A818543-250MG A818543-5G
77293-872EA 65.92 USD
77293-872 77618-292 77618-294 77618-296
4-(Ethoxymethylene)-2-phenyloxazol-5(4H)-one 95%
4-(Ethoxymethylene)-2-phenyloxazol-5(4H)-one

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₃
MW: 217.22 g/mol
MDL Number: MFCD00003204
CAS Number: 15646-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR