5-Fluoro-1H-imidazole 95%

Supplier: Ambeed
Warning

A818311-100MG A818311-250MG
77293-868EA 35 USD
77293-868 77293-870
5-Fluoro-1H-imidazole 95%
5-Fluoro-1H-imidazole

Precaución: for Research Use Only

Formula: C₃H₃FN₂
MDL Number: MFCD16295010
CAS Number: 30086-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR