2',3,4,5-Tetrafluoro-4''-propyl-1,1':4',1''-terphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 2',3,4,5-Tetrafluoro-4''-propyl-p-terphenyl

A817867-1G A817867-5G
77256-092EA 19.57 USD
77256-092 77256-094
2',3,4,5-Tetrafluoro-4''-propyl-1,1':4',1''-terphenyl 98%
2',3,4,5-Tetrafluoro-4''-propyl-1,1':4',1''-terphenyl

Precaución: for Research Use Only

Formula: C₂₁H₁₆F₄
MW: 344.35 g/mol
MDL Number: MFCD12964272
CAS Number: 205806-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR