3-Chloro-4-(ethylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-(ethylcarbamoyl)benzeneboronic acid, (3-Chloro-4-(ethylcarbamoyl)phenyl)boronic acid

A817207-1G A817207-250MG A817207-5G
77256-086EA 129.78 USD
77256-086 77256-088 77256-090
3-Chloro-4-(ethylcarbamoyl)phenylboronic acid 98%
3-Chloro-4-(ethylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BClNO₃
MW: 227.45 g/mol
MDL Number: MFCD07363767
CAS Number: 850589-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR