2,4-Dihydroxybenzamide 98%

Supplier: Ambeed
Warning

A817064-100G A817064-25G A817064-5G
77281-458EA 169 USD
77281-458 77281-460 77281-462
2,4-Dihydroxybenzamide 98%
2,4-Dihydroxybenzamide

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MDL Number: MFCD00064913
CAS Number: 3147-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR