3-Bromo-5-methylpyridin-4-ol 97%

Supplier: Ambeed
Warning

A816851-100MG A816851-1G A816851-250MG
77268-344EA 99.91 USD
77268-344 77268-346 77268-348
3-Bromo-5-methylpyridin-4-ol 97%
3-Bromo-5-methylpyridin-4-ol

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.02 g/mol
CAS Number: 1822926-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR