Neopentyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: 2,2-Dimethyl-1-propyl bromide, 1-Bromo-2,2-dimethylpropane

A816781-10G A816781-1G A816781-5G A816781-25G
77244-306EA 42 USD
77244-306 77244-308 77244-310 77552-510
Neopentyl bromide 98%
Neopentyl bromide

Precaución: for Research Use Only

Formula: C₅H₁₁Br
MDL Number: MFCD00000209
CAS Number: 630-17-1
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR