4,5-Difluoro-2-hydroxybenzonitrile 98%
Supplier: Ambeed
|
Danger
|
A816601-1G
A816601-100MG
77293-866EA
246.17
USD
77293-866
77749-604
4,5-Difluoro-2-hydroxybenzonitrile 98%
4,5-Difluoro-2-hydroxybenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₃F₂NO MW: 155.10 g/mol |
MDL Number:
MFCD08275287 CAS Number: 186590-36-3 UN: 3439 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...