3-Chloro-2-methoxybenzoic acid 96%

Supplier: Ambeed
Danger

A816270-10G A816270-1G A816270-250MG A816270-25G
77281-450EA 57 USD
77281-450 77281-452 77281-454 77281-456
3-Chloro-2-methoxybenzoic acid 96%
3-Chloro-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MDL Number: MFCD00179306
CAS Number: 3260-93-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR