3,5-Dibromo-4-methoxybenzoic acid 95%

Supplier: Ambeed
Warning

A815445-1G A815445-250MG A815445-5G
77281-440EA 17 USD
77281-440 77281-442 77281-444
3,5-Dibromo-4-methoxybenzoic acid 95%
3,5-Dibromo-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₃
MDL Number: MFCD00016511
CAS Number: 4073-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR