1H,1H,3H-Hexafluorobutyl acrylate 98% stabilized

Supplier: Ambeed
Danger

Synonyms: HFBA, 2,2,3,4,4,4-Hexafluorobutyl Acrylate, Acrylic acid 2,2,3,4,4,4-Hexafluorobutyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A815073-25G A815073-5G
77256-080EA 21.63 USD
77256-080 77256-082
1H,1H,3H-Hexafluorobutyl acrylate 98% stabilized
1H,1H,3H-Hexafluorobutyl acrylate

Precaución: for Research Use Only

Formula: C₇H₆F₆O₂
MW: 236.1118 g/mol
MDL Number: MFCD00044091
CAS Number: 54052-90-3
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR