3',4'-Dichloropropiophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3,4-Dichlorophenyl)-1-propanone, 1-(3,4-Dichlorophenyl)propan-1-one

A814173-1G A814173-25G A814173-5G
77281-434EA 14.42 USD
77281-434 77281-436 77281-438
3',4'-Dichloropropiophenone 98%
3',4'-Dichloropropiophenone

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O
MW: 203.07 g/mol
MDL Number: MFCD00000554
CAS Number: 6582-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR