3-Hydroxy-5-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A813663-1G A813663-5G
77281-432EA 16 USD
77281-432 77749-600
3-Hydroxy-5-nitrobenzoic acid 97%
3-Hydroxy-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₅
MDL Number: MFCD01098212
CAS Number: 78238-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR