1,10-Phenanthrolin-4-ol 97%

Supplier: Ambeed
Warning

A813417-1G A813417-250MG A813417-5G A813417-100MG
77244-294EA 47 USD
77244-294 77244-296 77618-278 77747-568
1,10-Phenanthrolin-4-ol 97%
1,10-Phenanthrolin-4-ol

Precaución: for Research Use Only

Formula: C₁₂H₈N₂O
MDL Number: MFCD00957343
CAS Number: 23443-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR