tert-Butyl (4R,6R)-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate 95%

Supplier: Ambeed
Warning

Synonyms: (4R,6R)-6-Cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetic Acid tert-Butyl Ester, tert-Butyl 2-((4R,6R)-6-(cyanomethyl)-2,2-dimethyl-1,3-dioxan-4-yl)acetate

A813304-100G A813304-5G A813304-25G
77244-290EA 125 USD
77244-290 77244-292 77747-567
tert-Butyl (4R,6R)-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate 95%
tert-Butyl (4R,6R)-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₄
MDL Number: MFCD03093958
CAS Number: 125971-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR