4-(Difluoromethoxy)benzonitrile 98%
Supplier: Ambeed
|
Danger
|
A813166-1G
A813166-25G
A813166-5G
77244-284EA
35
USD
77244-284
77244-286
77244-288
4-(Difluoromethoxy)benzonitrile 98%
4-(Difluoromethoxy)benzonitrile
Precaución: for Research Use Only
Formula:
C₈H₅F₂NO |
MDL Number:
MFCD00085006 CAS Number: 90446-25-6 UN: 3439 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...