(S)-(+)-1-[(R)-2-(Di-2-furylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (2S)-1-[(1S)-1-[Bis(3,5-dimethylphenyl)phosphino]ethyl]-2-(di-2-furanylphosphino)ferrocene

A811686-100MG A811686-250MG
77244-272EA 78.28 USD
77244-272 77244-274
(S)-(+)-1-[(R)-2-(Di-2-furylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine ≥98%, ee 99%
(S)-(+)-1-[(R)-2-(Di-2-furylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine

Precaución: for Research Use Only

Formula: C₃₆H₃₆FeO₂P₂
MW: 618.4700 g/mol
MDL Number: MFCD06658128
CAS Number: 649559-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR