1,4-Di[[2,2':6',2''-terpyridin]-4'-yl]benzene 97%

Supplier: Ambeed
Warning

Synonyms: 1,4-Di([2,2':6',2''-terpyridin]-4'-yl)benzene

A811628-100MG A811628-1G A811628-250MG A811628-5G
77293-848EA 26.78 USD
77293-848 77293-850 77293-852 77293-854
1,4-Di[[2,2':6',2''-terpyridin]-4'-yl]benzene 97%
1,4-Di[[2,2':6',2''-terpyridin]-4'-yl]benzene

Precaución: for Research Use Only

Formula: C₃₆H₂₄N₆
MW: 540.63 g/mol
MDL Number: MFCD02093699
CAS Number: 146406-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR