1-(4-Bromophenyl)-3-(dimethylamino)propan-1-one hydrochloride 97%

Supplier: Ambeed
Warning

A811286-100MG A811286-1G A811286-250MG
77256-064EA 12 USD
77256-064 77256-066 77256-068
1-(4-Bromophenyl)-3-(dimethylamino)propan-1-one hydrochloride 97%
1-(4-Bromophenyl)-3-(dimethylamino)propan-1-one hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrClNO
MDL Number: MFCD00514560
CAS Number: 881-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR