Copper(II) hexafluoroacetylacetonate hydrate 98%

Supplier: Ambeed
Warning

Synonyms: Hexafluoroacetylacetono Copper(II) Salt, Copper(II) Hexafluoroacetylacetonate, Copper(II) hexafluoro-2,4-pentanedionate hydrate, Bis(hexafluoroacetylacetonato)copper(II) hydrate, Copper(II) hexafluoro-2,4-pentanedionate xhydrate

A811084-1G A811084-5G A811084-25G
77256-060EA 24 USD
77256-060 77256-062 77618-274
Copper(II) hexafluoroacetylacetonate hydrate 98%
Copper (II) hexafluoroacetylacetonate hydrate

Precaución: for Research Use Only

Formula: C₁₀H₂CuF₁₂O₄·xH₂O
MW: 477.64 g/mol (anhy)
MDL Number: MFCD00151019
CAS Number: 155640-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR