2-Isopropylanisole 98%

Supplier: Ambeed
Warning

A810854-100G A810854-25G A810854-5G
77268-324EA 119 USD
77268-324 77268-326 77268-328
2-Isopropylanisole 98%
2-Isopropylanisole

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MDL Number: MFCD00798590
CAS Number: 2944-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR