6-Methoxy-2-(5-methylfuran-2-yl)-N-(1-methylpiperidin-4-yl)-7-(3-(pyrrolidin-1-yl)propoxy)quinolin-4-amine ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
CM-272
A810645-1MG
A810645-5MG
A810645-10MG
A810645-50MG
A810645-100MG
77268-320EA
28.84
USD
77268-320
77268-322
77618-268
77749-597
77749-596
6-Methoxy-2-(5-methylfuran-2-yl)-N-(1-methylpiperidin-4-yl)-7-(3-(pyrrolidin-1-yl)propoxy)quinolin-4-amine ≥98%
6-Methoxy-2-(5-methylfuran-2-yl)-N-(1-methylpiperidin-4-yl)-7-(3-(pyrrolidin-1-yl)propoxy)quinolin-4-amine
Precaución: for Research Use Only
Formula:
C₂₈H₃₈N₄O₃ MW: 478.63 g/mol |
MDL Number:
MFCD31812316 CAS Number: 1846570-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...