3,5-Dimethyl-1H-pyrrole-2,4-dicarboxylic acid 95%

Supplier: Ambeed
Warning

A810035-100MG A810035-1G A810035-250MG
77256-054EA 32.96 USD
77256-054 77256-056 77256-058
3,5-Dimethyl-1H-pyrrole-2,4-dicarboxylic acid 95%
3,5-Dimethyl-1H-pyrrole-2,4-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₄
MDL Number: MFCD00826671
CAS Number: 5434-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR