5-Amino-3-methylthiophene-2,4-dicarbonitrile 97%

Supplier: Ambeed
Warning

A809952-1G A809952-250MG
77256-050EA 30 USD
77256-050 77256-052
5-Amino-3-methylthiophene-2,4-dicarbonitrile 97%
5-Amino-3-methylthiophene-2,4-dicarbonitrile

Precaución: for Research Use Only

Formula: C₇H₅N₃S
MDL Number: MFCD01105451
CAS Number: 52603-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR