(±)-3-Ethoxy-1,2-propanediol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(±)-3-Ethoxy-1,2-dihydroxy propane, 3-Ethoxy-1,2-propanediol
A809867-100MG
A809867-1G
A809867-250MG
A809867-5G
77281-414EA
5.15
USD
77281-414
77281-416
77281-418
77281-420
(±)-3-Ethoxy-1,2-propanediol 95%
(±)-3-Ethoxy-1,2-propanediol
Precaución: for Research Use Only
Formula:
C₅H₁₂O₃ MW: 120.15 g/mol |
MDL Number:
MFCD00009989 CAS Number: 1874-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...