tert-Butyl 4-(3-aminophenyl)piperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

A809779-100MG A809779-1G A809779-250MG A809779-5G
77293-832EA 19.57 USD
77293-832 77293-834 77293-836 77293-838
tert-Butyl 4-(3-aminophenyl)piperazine-1-carboxylate 98%
tert-Butyl 4-(3-aminophenyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₃N₃O₂
MDL Number: MFCD06657283
CAS Number: 206879-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR