Ethyl 2-chloro-5-methylisonicotinate 97%

Supplier: Ambeed
Warning

A809420-100MG A809420-1G
77268-316EA 64.89 USD
77268-316 77618-264
Ethyl 2-chloro-5-methylisonicotinate 97%
Ethyl 2-chloro-5-methylisonicotinate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.63 g/mol
MDL Number: MFCD06410741
CAS Number: 850080-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR