3,3,5-Trimethylcaproic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,3,5-Trimethylhexanoic acid, 3,5,5-Trimethylhexanoicacid, 3,5,5-Trimethylhexanoic acid

A808927-100G A808927-500G
77281-410EA 6.18 USD
77281-410 77281-412
3,3,5-Trimethylcaproic acid 98%
3,3,5-Trimethylcaproic acid

Precaución: for Research Use Only

Formula: C₉H₁₈O₂
MW: 158.24 g/mol
MDL Number: MFCD00020507
CAS Number: 3302-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR