2-Ethoxybenzylamine 98%

Supplier: Ambeed
Warning

A808303-1G A808303-250MG A808303-5G
77268-302EA 32 USD
77268-302 77268-304 77268-306
2-Ethoxybenzylamine 98%
2-Ethoxybenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MDL Number: MFCD00008111
CAS Number: 37806-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR