1,4-Bis(4-amino-2-trifluoromethylphenoxy)benzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,4'-(1,4-Phenylenebis(oxy))bis(3-(trifluoromethyl)aniline)
A807912-100G
A807912-1G
A807912-25G
A807912-5G
77293-824EA
276.04
USD
77293-824
77293-826
77293-828
77293-830
1,4-Bis(4-amino-2-trifluoromethylphenoxy)benzene 98%
1,4-Bis(4-amino-2-trifluoromethylphenoxy)benzene
Precaución: for Research Use Only
Formula:
C₂₀H₁₄F₆N₂O₂ MW: 428.33 g/mol |
MDL Number:
MFCD07368071 CAS Number: 94525-05-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...