Ethyl 4-(2-hydroxypropan-2-yl)-2-propyl-1-((2'-(1-trityl-1H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-imidazole-5-carboxylate 97%

Supplier: Ambeed
Danger

A807875-10G A807875-1G A807875-250MG A807875-5G
77256-036EA 36.05 USD
77256-036 77256-038 77256-040 77256-042
Ethyl 4-(2-hydroxypropan-2-yl)-2-propyl-1-((2'-(1-trityl-1H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-imidazole-5-carboxylate 97%
Ethyl 4-(2-hydroxypropan-2-yl)-2-propyl-1-((2'-(1-trityl-1H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-imidazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₄₅H₄₄N₆O₃
MW: 716.86 g/mol
MDL Number: MFCD11973634
CAS Number: 144690-33-5
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR