2,2-Bis(4-hydroxycyclohexyl)propane (mixture of isomeres) 98%

Supplier: Ambeed
Warning

A807821-1000G A807821-100G A807821-25G A807821-500G
77256-028EA 85 USD
77256-028 77256-030 77256-032 77256-034
2,2-Bis(4-hydroxycyclohexyl)propane (mixture of isomeres) 98%
2,2-Bis(4-hydroxycyclohexyl)propane (mixture of isomeres)

Precaución: for Research Use Only

Formula: C₁₅H₂₈O₂
MDL Number: MFCD00019334
CAS Number: 80-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR