(R,R)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline) ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (4R,4'R)-2,2'-(Propane-2,2-diyl)bis(4-isopropyl-4,5-dihydrooxazole), (R,R)-2,2-Bis(4-isopropyl-2-oxazolin-2-yl)propane, 2,2-Bis[(4R)-4-isopropyl-2-oxazolin-2-yl]propane

A807780-100MG A807780-1G A807780-250MG A807780-50MG
77256-020EA 28.84 USD
77256-020 77256-022 77256-024 77256-026
(R,R)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline) ≥98%, ee 99%
(R,R)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline)

Precaución: for Research Use Only

Formula: C₁₅H₂₆N₂O₂
MW: 266.39 g/mol
MDL Number: MFCD28038687
CAS Number: 150529-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR