2,4-Bis(4-biphenylyl)-6-chloro-1,3,5-triazine 97%

Supplier: Ambeed
Warning

Synonyms: 2,4-Di([1,1'-biphenyl]-4-yl)-6-chloro-1,3,5-triazine

A807744-1G A807744-25G A807744-5G
77244-256EA 6 USD
77244-256 77244-258 77244-260
2,4-Bis(4-biphenylyl)-6-chloro-1,3,5-triazine 97%
2,4-Bis(4-biphenylyl)-6-chloro-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₂₇H₁₈ClN₃
MW: 419.91 g/mol
MDL Number: MFCD27979765
CAS Number: 182918-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR