1-Undecanol 99%

Supplier: Ambeed
Danger

Synonyms: 1-Undecyl alcohol, Undecyl alcohol, Undecan-1-ol, Decyl carbinol, 1-Hydroxy undecane

A807663-1G A807663-5G A807663-25G A807663-100G A807663-500G
77244-252EA 5.15 USD
77244-252 77244-254 77552-490 77618-256 77747-554
1-Undecanol 99%
1-Undecanol

Precaución: for Research Use Only

Formula: C₁₁H₂₄O
MW: 172.31 g/mol
MDL Number: MFCD00004751
CAS Number: 112-42-5
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR