Methanesulfonato{(R)-(-)-1-[(S)-2-(dicyclohexylphosphino)ferrocenyl]ethyldi-t-butylphosphine}(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%

Supplier: Ambeed
Warning

A807416-100MG A807416-1G A807416-250MG A807416-5G
77256-014EA 21 USD
77256-014 77256-016 77256-018 77552-486
Methanesulfonato{(R)-(-)-1-[(S)-2-(dicyclohexylphosphino)ferrocenyl]ethyldi-t-butylphosphine}(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%
Methanesulfonato{(R)-(-)-1-[(S)-2-(dicyclohexylphosphino)ferrocenyl]ethyldi-t-butylphosphine}(2'-amino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: for Research Use Only

Formula: C₄₅H₆₅FeNO₃P₂PdS
MDL Number: MFCD27978424
CAS Number: 1702311-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR