(R)-tert-Butyl (1-(7-(but-2-yn-1-yl)-3-methyl-1-((4-methylquinazolin-2-yl)methyl)-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-8-yl)piperidin-3-yl)carbamate 97% (by HPLC)

Supplier: Ambeed
Danger

A807245-1G A807245-5G
77256-010EA 39.14 USD
77256-010 77256-012
(R)-tert-Butyl (1-(7-(but-2-yn-1-yl)-3-methyl-1-((4-methylquinazolin-2-yl)methyl)-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-8-yl)piperidin-3-yl)carbamate 97% (by HPLC)
(R)-tert-Butyl (1-(7-(but-2-yn-1-yl)-3-methyl-1-((4-methylquinazolin-2-yl)methyl)-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-8-yl)piperidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₃₀H₃₆N₈O₄
MW: 572.65 g/mol
MDL Number: MFCD30489588
CAS Number: 668273-75-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR