2,2-Difluoro-4-pentenoic acid 95%

Supplier: Ambeed
Danger

A807092-100MG A807092-1G A807092-250MG A807092-25G A807092-5G
77256-000EA 10 USD
77256-000 77256-002 77256-004 77256-006 77256-008
2,2-Difluoro-4-pentenoic acid 95%
2,2-Difluoro-4-pentenoic acid

Precaución: for Research Use Only

Formula: C₅H₆F₂O₂
MDL Number: MFCD09800642
CAS Number: 55039-89-9
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR