2-(2-Acetamidophenoxy)acetic acid 97%

Supplier: Ambeed
Warning

A806339-10G A806339-250MG A806339-5G
77244-236EA 92 USD
77244-236 77244-238 77244-240
2-(2-Acetamidophenoxy)acetic acid 97%
2-(2-Acetamidophenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₄
MDL Number: MFCD00455437
CAS Number: 1798-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR