4-Biphenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

A806030-100G A806030-10G A806030-1G A806030-25G A806030-5G
77293-812EA 251.32 USD
77293-812 77293-814 77293-816 77293-818 77293-820
4-Biphenylboronic acid pinacol ester 98%
4-Biphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₁BO₂
MW: 280.17 g/mol
MDL Number: MFCD07368291
CAS Number: 144432-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR