2',3',4',5',6'-Pentafluoroacetophenone 98%

Supplier: Ambeed
Warning

A805859-100G A805859-1G A805859-25G A805859-5G
77255-992EA 645.81 USD
77255-992 77255-994 77255-996 77255-998
2',3',4',5',6'-Pentafluoroacetophenone 98%
2',3',4',5',6'-Pentafluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₃F₅O
MW: 210.1 g/mol
MDL Number: MFCD00000296
CAS Number: 652-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR