3-(Cyanomethyl)-2,4,5-trimethylthiophene 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2,4,5-Trimethylthiophen-3-yl)acetonitrile

A804929-100MG A804929-250MG
77255-988EA 24.72 USD
77255-988 77255-990
3-(Cyanomethyl)-2,4,5-trimethylthiophene 98%
3-(Cyanomethyl)-2,4,5-trimethylthiophene

Precaución: for Research Use Only

Formula: C₉H₁₁NS
MW: 165.25 g/mol
MDL Number: MFCD00143113
CAS Number: 112440-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR