Propargyl-PEG4-acid 97%

Supplier: Ambeed
Warning

Synonyms: 4,7,10,13-Tetraoxahexadec-15-yn-1-oic acid

A804377-100MG A804377-1G A804377-5G A804377-250MG
77268-282EA 16 USD
77268-282 77268-284 77268-286 77618-252
Propargyl-PEG4-acid 97%
Propargyl-PEG4-acid

Precaución: for Research Use Only

Formula: C₁₂H₂₀O₆
MDL Number: MFCD22683284
CAS Number: 1415800-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR