2-Chloro-4-fluorobenzyl alcohol 98%

Supplier: Ambeed
Warning

A803959-100G A803959-25G A803959-5G
77268-276EA 228.66 USD
77268-276 77268-278 77268-280
2-Chloro-4-fluorobenzyl alcohol 98%
2-Chloro-4-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆ClFO
MW: 160.58 g/mol
MDL Number: MFCD00236024
CAS Number: 208186-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR