3,3,3-Trifluoro-1-propanol 95%

Supplier: Ambeed
Danger

Synonyms: 3,3,3-Trifluoropropan-1-ol

A803777-1G A803777-25G A803777-5G
77281-402EA 9 USD
77281-402 77281-404 77281-406
3,3,3-Trifluoro-1-propanol 95%
3,3,3-Trifluoro-1-propanol

Precaución: for Research Use Only

Formula: C₃H₅F₃O
MW: 114.07 g/mol
MDL Number: MFCD00190649
CAS Number: 2240-88-2
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR