5-Chloro-(2-ethoxycarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-2-(ethoxycarbonyl)benzeneboronic acid, (5-Chloro-2-(ethoxycarbonyl)phenyl)boronic acid

A803688-1G A803688-250MG
77255-982EA 109 USD
77255-982 77255-984
5-Chloro-(2-ethoxycarbonyl)phenylboronic acid 98%
5-Chloro-(2-ethoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BClO₄
MDL Number: MFCD07363744
CAS Number: 871329-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR