N,N'-Bis[4-(diphenylamino)phenyl]-N,N'-di(1-naphthyl)benzidine 98%

Supplier: Ambeed
Warning

Synonyms: N1,N1'-([1,1'-Biphenyl]-4,4'-diyl)bis(N1-(naphthalen-1-yl)-N4,N4-diphenylbenzene-1,4-diamine)

A803581-100MG A803581-1G A803581-250MG
77255-976EA 20.6 USD
77255-976 77255-978 77255-980
N,N'-Bis[4-(diphenylamino)phenyl]-N,N'-di(1-naphthyl)benzidine 98%
N,N'-Bis[4-(diphenylamino)phenyl]-N,N'-di(1-naphthyl)benzidine

Precaución: for Research Use Only

Formula: C₆₈H₅₀N₄
MW: 923.18 g/mol
MDL Number: MFCD20488037
CAS Number: 910058-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR